Assay Details
Target Gene Details
Entrez Gene ID: | 814 |
Gene Name: | calcium/calmodulin dependent protein kinase IV |
Gene Aliases: |
CaMK IV, CaMK-GR, caMK |
Location: |
Chr.5:111223653-111498503 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK4 | NM_001323374.1 | 2 | 189 | NP_001310303.1 |
| NM_001323375.1 | 2 | 221 | NP_001310304.1 | |
| NM_001323376.1 | 1 | 337 | NP_001310305.1 | |
| NM_001323377.1 | NP_001310306.1 | |||
| NM_001744.5 | 1 | 337 | NP_001735.1 | |
| AK311662.1 | 1 | 192 | ||
| AK312840.1 | 1 | 192 | ||
| BC016695.1 | 1 | 181 | AAH16695.2 | |
| BC025687.1 | 1 | 179 | AAH25687.1 | |
| BG721313.1 | 1 | 195 | ||
| D30742.1 | 1 | 167 | BAA06403.1 | |
| DA114495.1 | 1 | 337 | ||
| DA396146.1 | 1 | 325 | ||
| L17000.1 | 1 | 131 | AAA35639.1 | |
| L24959.1 | 2 | 189 | AAA18251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606259 | Chr.5:110985051 - 111262481 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map