Assay Details
Target Gene Details
| Entrez Gene ID: | 388611 | 
| Gene Name: | CD164 molecule like 2 | 
| Gene Aliases: | EAPG6122, UNQ6122 | 
| Location: | Chr.1:27379100-27383333 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Intron 2 - Exon 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CD164L2 | NM_207397.3 | NP_997280.2 | ||
| XM_005245868.3 | XP_005245925.2 | |||
| XM_011541441.1 | XP_011539743.1 | |||
| AY358761.1 | AAQ89121.1 | |||
| BC137466.1 | ||||
| DB114521.1 | ||||
| DB231017.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv526051 | Chr.1:27284255 - 27445496 on Build GRCh38 | Gain |  SYTL1  FCN3  WASF2  GPR3  WDTC1  CD164L2  MAP3K6  ACTG1P20  TMEM222 | 
| nsv516766 | Chr.1:27335744 - 27405147 on Build GRCh38 | Loss |  SYTL1  FCN3  WASF2  GPR3  CD164L2  MAP3K6  TMEM222 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic