Assay Details
Target Gene Details
Entrez Gene ID: | 81888 |
Gene Name: | hydroxypyruvate isomerase (putative) |
Gene Aliases: |
HT036 |
Location: |
Chr.1:43450607-43454241 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HYI | XM_005271239.4 | 1 | 158 | XP_005271296.1 |
| XM_005271240.4 | 1 | 158 | XP_005271297.1 | |
| XM_006710937.3 | 1 | 158 | XP_006711000.1 | |
| XM_011542222.2 | 1 | 158 | XP_011540524.1 | |
| AF284750.1 | 1 | 170 | AAG59852.1 | |
| AF284751.1 | 1 | 170 | AAG59853.1 |
Target Gene Details
Entrez Gene ID: | 23334 |
Gene Name: | seizure threshold 2 homolog (mouse) |
Gene Aliases: |
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B |
Location: |
Chr.1:43389884-43454247 on Build GRCh38 |
Assay Gene Location: | Within Exon 75 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SZT2 | NM_015284.3 | 71 | 13816 | NP_056099.3 |
| BU633294.1 | 1 | 181 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950702 | Chr.1:43454030 - 43457429 on Build GRCh38 | Duplication |
|
| nsv438377 | Chr.1:43451738 - 43468018 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map