Assay Details
Target Gene Details
Entrez Gene ID: | 405 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator |
Gene Aliases: |
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2 |
Location: |
Chr.1:150809705-150876768 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437169 | Chr.1:150765721 - 150844410 on Build GRCh38 | Loss |
|
| nsv2788 | Chr.1:150782858 - 150828309 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map