Assay Details
Target Gene Details
Entrez Gene ID: | 163 |
Gene Name: | adaptor related protein complex 2 beta 1 subunit |
Gene Aliases: |
ADTB2, AP105B, AP2-BETA, CLAPB1 |
Location: |
Chr.17:35586899-35726417 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP2B1 | NM_001030006.1 | NP_001025177.1 | ||
| NM_001282.2 | NP_001273.1 | |||
| XM_005257941.2 | XP_005257998.1 | |||
| XM_011524448.1 | 1 | 385 | XP_011522750.1 | |
| XM_011524451.1 | XP_011522753.1 | |||
| XM_011524452.1 | XP_011522754.1 | |||
| XM_011524453.1 | XP_011522755.1 | |||
| XM_011524455.2 | XP_011522757.1 | |||
| XM_017024285.1 | XP_016879774.1 | |||
| AK289528.1 | ||||
| AK295667.1 | ||||
| AK301522.1 | ||||
| AY341427.1 | AAQ20044.1 | |||
| BX643620.1 | ||||
| M34175.1 | AAA35583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2038 | Chr.17:35569617 - 35598796 on Build GRCh38 | Insertion |
|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map