Assay Details
Target Gene Details
Entrez Gene ID: | 23584 |
Gene Name: | V-set and immunoglobulin domain containing 2 |
Gene Aliases: |
2210413P10Rik, CTH, CTXL |
Location: |
Chr.11:124747474-124752213 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSIG2 | NM_014312.3 | 2 | 173 | NP_055127.2 |
| AK000460.1 | 2 | 154 | BAA91179.1 | |
| AY358897.1 | 2 | 198 | AAQ89256.1 | |
| BC007313.2 | 2 | 173 | AAH07313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627986 | Chr.11:124740300 - 124752449 on Build GRCh38 | Gain |
|
| nsv556487 | Chr.11:124730862 - 124765248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map