Assay Details
Target Gene Details
Entrez Gene ID: | 3383 |
Gene Name: | intercellular adhesion molecule 1 |
Gene Aliases: |
BB2, CD54, P3.58 |
Location: |
Chr.19:10270841-10286615 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICAM1 | NM_000201.2 | 3 | 840 | NP_000192.2 |
| AF340038.1 | 3 | 521 | AAQ14901.1 | |
| AF340039.1 | 3 | 509 | AAQ14902.1 | |
| AK130659.1 | 3 | 561 | ||
| AK298054.1 | 3 | 447 | ||
| AK298983.1 | 3 | 273 | ||
| AK301412.1 | ||||
| AK312636.1 | 3 | 561 | ||
| BC015969.2 | 3 | 592 | AAH15969.1 | |
| BT006854.1 | 3 | 521 | AAP35500.1 | |
| J03132.1 | 3 | 578 | AAA52709.1 | |
| M24283.1 | 3 | 592 | AAA52708.1 | |
| M55024.1 | 2 | 268 | AAA60392.1 | |
| X06990.1 | 3 | 533 | CAA30051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578612 | Chr.19:10271961 - 10284355 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map