Assay Details
Target Gene Details
Entrez Gene ID: | 405 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator |
Gene Aliases: |
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2 |
Location: |
Chr.1:150809705-150876768 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNT | NM_001197325.1 | 21 | 4057 | NP_001184254.1 |
| NM_001286035.1 | 23 | 4146 | NP_001272964.1 | |
| NM_001286036.1 | 22 | 4099 | NP_001272965.1 | |
| NM_001668.3 | 22 | 4105 | NP_001659.1 | |
| NM_178427.2 | 21 | 4060 | NP_848514.1 | |
| XM_005245151.1 | 22 | 4044 | XP_005245208.1 | |
| XM_005245153.1 | 22 | 4032 | XP_005245210.1 | |
| XM_005245154.3 | 23 | 4075 | XP_005245211.1 | |
| XM_005245157.1 | 21 | 3936 | XP_005245214.1 | |
| XM_011509542.2 | 22 | 4108 | XP_011507844.1 | |
| XM_011509543.2 | 23 | 4103 | XP_011507845.1 | |
| XM_011509544.2 | 22 | 4072 | XP_011507846.1 | |
| XM_011509545.2 | 21 | 4063 | XP_011507847.1 | |
| XM_011509546.1 | 21 | 3823 | XP_011507848.1 | |
| XM_011509547.2 | 21 | 4060 | XP_011507849.1 | |
| XM_017001288.1 | 23 | 3947 | XP_016856777.1 | |
| XM_017001289.1 | 23 | 3935 | XP_016856778.1 | |
| XM_017001290.1 | 22 | 3904 | XP_016856779.1 | |
| XM_017001291.1 | 22 | 3903 | XP_016856780.1 | |
| XM_017001292.1 | 22 | 3840 | XP_016856781.1 | |
| XM_017001293.1 | 21 | 3794 | XP_016856782.1 | |
| XM_017001294.1 | 21 | 3933 | XP_016856783.1 | |
| XM_017001295.1 | 21 | 3921 | XP_016856784.1 | |
| XM_017001296.1 | 20 | 3900 | XP_016856785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437169 | Chr.1:150765721 - 150844410 on Build GRCh38 | Loss |
|
| nsv2788 | Chr.1:150782858 - 150828309 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map