Assay Details
Target Gene Details
Entrez Gene ID: | 10240 |
Gene Name: | mitochondrial ribosomal protein S31 |
Gene Aliases: |
IMOGN38, MRP-S31, S31mt |
Location: |
Chr.13:40729296-40771220 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS31 | NM_005830.3 | NP_005821.2 | ||
| AK315248.1 | ||||
| AV712217.1 | ||||
| BC022045.1 | AAH22045.1 | |||
| Z68747.1 | CAA92951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892332 | Chr.13:40619338 - 40820761 on Build GRCh38 | Gain |
|
| nsv974239 | Chr.13:40724480 - 40774465 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map