Assay Details
Target Gene Details
Entrez Gene ID: | 100127889 |
Gene Name: | chromosome 10 open reading frame 131 |
Gene Aliases: |
bA690P14.3 |
Location: |
Chr.10:95907965-95938658 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf131 | NM_001130446.2 | NP_001123918.2 | ||
| AK294399.1 |
Target Gene Details
Entrez Gene ID: | 728558 |
Gene Name: | ENTPD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:95753206-96090235 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD1-AS1 | NR_038444.1 | |||
| AK091396.1 | ||||
| AK096469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv25405 | Chr.10:95923269 - 95926815 on Build GRCh38 | Deletion |
|

Genomic Map