Assay Details
Target Gene Details
Entrez Gene ID: | 119180 |
Gene Name: | lysozyme like 2 |
Gene Aliases: |
LYZD2 |
Location: |
Chr.10:30606222-30629761 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYZL2 | NM_183058.2 | NP_898881.2 | ||
| XM_011519306.2 | XP_011517608.1 | |||
| AF139543.1 | AAP97272.1 | |||
| AI637906.1 | ||||
| BC066294.1 | AAH66294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831822 | Chr.10:30459326 - 30628314 on Build GRCh38 | Gain |
|
| nsv1161936 | Chr.10:30113251 - 31078102 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map