Assay Details
Target Gene Details
Entrez Gene ID: | 728192 |
Gene Name: | long intergenic non-protein coding RNA 460 |
Gene Aliases: |
- |
Location: |
Chr.13:106376563-106377792 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00460 | NR_034119.1 | |||
| BI544279.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633458 | Chr.13:106127915 - 106528334 on Build GRCh38 | Gain |
|
| nsv1039628 | Chr.13:106180176 - 106649540 on Build GRCh38 | Gain |
|

Genomic Map