Assay Details
Target Gene Details
Entrez Gene ID: | 80781 |
Gene Name: | collagen type XVIII alpha 1 chain |
Gene Aliases: |
KNO, KNO1, KS |
Location: |
Chr.21:45405137-45513720 on Build GRCh38 |
Assay Gene Location: | Within Intron 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL18A1 | NM_030582.3 | NP_085059.2 | ||
| NM_130444.2 | NP_569711.2 | |||
| NM_130445.3 | NP_569712.2 | |||
| AF018081.1 | AAC39658.1 | |||
| AF018082.1 | AAC39659.1 | |||
| AK098216.1 | ||||
| AK130835.1 | ||||
| BC033715.1 | AAH33715.1 | |||
| BC063833.1 | AAH63833.1 | |||
| L22548.1 | AAA51864.1 |
Target Gene Details
Entrez Gene ID: | 6573 |
Gene Name: | solute carrier family 19 member 1 |
Gene Aliases: |
CHMD, FOLT, IFC1, REFC, RFC1 |
Location: |
Chr.21:45502517-45563143 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC19A1 | XM_017028445.1 | XP_016883934.1 | ||
| AB209069.1 | BAD92306.1 | |||
| AK310797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516407 | Chr.21:45410958 - 45620742 on Build GRCh38 | Loss |
|
| esv2762114 | Chr.21:45260287 - 45647830 on Build GRCh38 | Gain |
|
| nsv953653 | Chr.21:45409686 - 45560386 on Build GRCh38 | Deletion |
|
| esv3893419 | Chr.21:45463239 - 45625553 on Build GRCh38 | Gain |
|
| dgv4437n100 | Chr.21:45453955 - 45546266 on Build GRCh38 | Gain |
|
| nsv834111 | Chr.21:45423171 - 45585330 on Build GRCh38 | Loss |
|
| nsv587853 | Chr.21:45506179 - 45562421 on Build GRCh38 | Loss |
|
| nsv587856 | Chr.21:45506637 - 45525426 on Build GRCh38 | Loss |
|
| nsv1061760 | Chr.21:45466796 - 45511534 on Build GRCh38 | Gain |
|
| dgv7903n54 | Chr.21:45496498 - 45540367 on Build GRCh38 | Loss |
|
| dgv4438n100 | Chr.21:45462733 - 45559862 on Build GRCh38 | Gain |
|
| dgv4436n100 | Chr.21:45402338 - 45556768 on Build GRCh38 | Gain |
|
| nsv834112 | Chr.21:45502237 - 45628339 on Build GRCh38 | Gain |
|
| nsv587851 | Chr.21:45495128 - 45517044 on Build GRCh38 | Loss |
|
| nsv587854 | Chr.21:45506383 - 45512201 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114834044] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map