Assay Details
Target Gene Details
Entrez Gene ID: | 10056 |
Gene Name: | phenylalanyl-tRNA synthetase beta subunit |
Gene Aliases: |
FARSLB, FRSB, HSPC173, PheHB, PheRS |
Location: |
Chr.2:222571443-222656355 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARSB | NM_005687.4 | NP_005678.3 | ||
| NR_130154.1 | ||||
| XM_006712169.2 | XP_006712232.1 | |||
| XM_011510466.2 | XP_011508768.1 | |||
| XM_017003110.1 | XP_016858599.1 | |||
| AF042346.1 | AAD02220.1 | |||
| AF161521.1 | AAF29136.1 | |||
| AK055289.1 | ||||
| AK291531.1 | ||||
| AK294158.1 | ||||
| AK316497.1 | ||||
| BC006502.2 | AAH06502.2 | |||
| BC017783.1 | AAH17783.1 | |||
| D84430.1 | BAA95608.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs181312534] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map