Assay Details
Target Gene Details
Entrez Gene ID: | 4045 |
Gene Name: | limbic system-associated membrane protein |
Gene Aliases: |
IGLON3, LAMP |
Location: |
Chr.3:115802363-116445542 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSAMP | NM_001318915.1 | NP_001305844.1 | ||
| NM_002338.4 | NP_002329.2 | |||
| XM_011512840.2 | XP_011511142.1 | |||
| XM_017006383.1 | XP_016861872.1 | |||
| XM_017006384.1 | XP_016861873.1 | |||
| AK315286.1 | ||||
| BC033803.1 | AAH33803.1 | |||
| BX647284.1 | 1 | 809 | ||
| U41901.1 | AAC50569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760762 | Chr.3:115757375 - 116088803 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map