Assay Details
Target Gene Details
Entrez Gene ID: | 1977 |
Gene Name: | eukaryotic translation initiation factor 4E |
Gene Aliases: |
AUTS19, CBP, EIF4E1, EIF4EL1, EIF4F, eIF-4E |
Location: |
Chr.4:98878456-98930635 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4E | NM_001130678.1 | NP_001124150.1 | ||
| NM_001130679.1 | NP_001124151.1 | |||
| NM_001968.3 | NP_001959.1 | |||
| XM_006714126.3 | XP_006714189.1 | |||
| AK295083.1 | ||||
| AK300982.1 | ||||
| AV708571.1 | ||||
| BC012611.1 | AAH12611.1 | |||
| BC035166.1 | AAH35166.1 | |||
| BC043226.1 | AAH43226.1 | |||
| BC107715.1 | AAI07716.1 | |||
| BM849222.1 | ||||
| DC388666.1 | ||||
| KJ534830.1 | ||||
| M15353.1 | AAC13647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27868 | Chr.4:98892383 - 98897090 on Build GRCh38 | Loss |
|
| esv3303064 | Chr.4:98892726 - 98895943 on Build GRCh38 | Tandem Duplication |
|
| nsv821288 | Chr.4:98891942 - 98897835 on Build GRCh38 | Duplication |
|
| esv1000667 | Chr.4:98892507 - 98895886 on Build GRCh38 | Gain |
|
| esv997164 | Chr.4:98893686 - 98895795 on Build GRCh38 | Insertion |
|
| esv2861680 | Chr.4:98892636 - 98895948 on Build GRCh38 | Duplication |
|
| nsv594943 | Chr.4:98893976 - 98895600 on Build GRCh38 | Gain |
|
| dgv1018e214 | Chr.4:98892259 - 98896019 on Build GRCh38 | Gain |
|
| nsv819967 | Chr.4:98892358 - 98896690 on Build GRCh38 | Loss |
|
| nsv956318 | Chr.4:98892940 - 98895632 on Build GRCh38 | Deletion |
|
| nsv950363 | Chr.4:98892750 - 98895949 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map