Assay Details
Target Gene Details
Entrez Gene ID: | 1977 |
Gene Name: | eukaryotic translation initiation factor 4E |
Gene Aliases: |
AUTS19, CBP, EIF4E1, EIF4EL1, EIF4F, eIF-4E |
Location: |
Chr.4:98878456-98930635 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4E | NM_001130678.1 | NP_001124150.1 | ||
| NM_001130679.1 | NP_001124151.1 | |||
| NM_001968.3 | NP_001959.1 | |||
| XM_006714126.3 | XP_006714189.1 | |||
| AK295083.1 | ||||
| AK300982.1 | ||||
| AV708571.1 | ||||
| BC012611.1 | AAH12611.1 | |||
| BC035166.1 | AAH35166.1 | |||
| BC043226.1 | AAH43226.1 | |||
| BC107715.1 | AAI07716.1 | |||
| BM849222.1 | ||||
| DC388666.1 | ||||
| KJ534830.1 | ||||
| M15353.1 | AAC13647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822666 | Chr.4:98891942 - 98895886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map