Assay Details
Target Gene Details
Entrez Gene ID: | 6462 |
Gene Name: | sex hormone binding globulin |
Gene Aliases: |
ABP, SBP, TEBG |
Location: |
Chr.17:7614064-7633383 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHBG | NM_001289114.1 | NP_001276043.1 | ||
| EU352670.1 | ||||
| EU352671.1 | ||||
| EU352672.1 | ||||
| EU352673.1 | ||||
| EU352674.1 | ||||
| EU352675.1 | ||||
| EU352676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map