Assay Details
Target Gene Details
Entrez Gene ID: | 4242 |
Gene Name: | MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase |
Gene Aliases: |
- |
Location: |
Chr.22:37469063-37486440 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFNG | NM_001166343.1 | NP_001159815.1 | ||
| NM_002405.3 | NP_002396.2 | |||
| NR_029413.1 | ||||
| AK292123.1 | ||||
| AK297149.1 | ||||
| AK297833.1 | ||||
| AK316555.1 | ||||
| BC094814.1 | AAH94814.1 | |||
| CR456518.1 | CAG30404.1 | |||
| U94352.1 | AAC51358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| nsv1057617 | Chr.22:37462633 - 37713698 on Build GRCh38 | Gain |
|
| esv3647709 | Chr.22:37466230 - 37470820 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map