Assay Details
Target Gene Details
Entrez Gene ID: | 2742 |
Gene Name: | glycine receptor alpha 2 |
Gene Aliases: |
GLR |
Location: |
Chr.X:14448779-14731812 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLRA2 | XM_011545495.2 | XP_011543797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528635 | Chr.X:14087371 - 14467523 on Build GRCh38 | Gain |
|
| nsv526475 | Chr.X:14363314 - 14702502 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map