Assay Details
Target Gene Details
| Entrez Gene ID: | 814 | 
| Gene Name: | calcium/calmodulin dependent protein kinase IV | 
| Gene Aliases: | CaMK IV, CaMK-GR, caMK | 
| Location: | Chr.5:111223653-111498503 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CAMK4 | NM_001323374.1 | NP_001310303.1 | ||
| NM_001323375.1 | NP_001310304.1 | |||
| NM_001323376.1 | NP_001310305.1 | |||
| NM_001323377.1 | NP_001310306.1 | |||
| NM_001744.5 | NP_001735.1 | |||
| AK311662.1 | ||||
| AK312840.1 | ||||
| BC016695.1 | AAH16695.2 | |||
| BC025687.1 | AAH25687.1 | |||
| BG721313.1 | ||||
| D30742.1 | BAA06403.1 | |||
| DA114495.1 | ||||
| DA396146.1 | ||||
| L17000.1 | AAA35639.1 | |||
| L24959.1 | AAA18251.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3606259 | Chr.5:110985051 - 111262481 on Build GRCh38 | Loss |  TSLP  CAMK4  WDR36 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top