Assay Details
Target Gene Details
Entrez Gene ID: | 11059 |
Gene Name: | WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
AIP5, Tiul1, hSDRP1 |
Location: |
Chr.8:86342711-86468357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WWP1 | NM_007013.3 | NP_008944.1 | ||
XM_005250760.3 | XP_005250817.1 | |||
XM_005250761.3 | XP_005250818.1 | |||
XM_017012993.1 | XP_016868482.1 | |||
XM_017012994.1 | XP_016868483.1 | |||
XM_017012995.1 | XP_016868484.1 | |||
XM_017012996.1 | XP_016868485.1 | |||
XM_017012997.1 | XP_016868486.1 | |||
AK292800.1 | ||||
AY345857.1 | AAQ22764.1 | |||
BC036065.1 | AAH36065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1017535 | Chr.8:86249473 - 86355705 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
