Assay Details
Target Gene Details
Entrez Gene ID: | 24 |
Gene Name: | ATP binding cassette subfamily A member 4 |
Gene Aliases: |
ABC10, ABCR, ARMD2, CORD3, FFM, RMP, RP19, STGD, STGD1 |
Location: |
Chr.1:93992837-94121149 on Build GRCh38 |
Assay Gene Location: | Within Intron 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCA4 | NM_000350.2 | NP_000341.2 | ||
| AB210040.1 | ||||
| AF000148.1 | AAC23915.1 | |||
| AF001945.1 | AAC05632.1 | |||
| AK296896.1 | ||||
| AK301733.1 | ||||
| AK301764.1 | ||||
| BM686882.1 | ||||
| CR627391.1 | CAH10486.1 | |||
| U66691.1 | ||||
| U88667.1 | AAC51144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516137 | Chr.1:93938682 - 94038989 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map