Assay Details
Target Gene Details
Entrez Gene ID: | 285203 |
Gene Name: | EGF domain specific O-linked N-acetylglucosamine transferase |
Gene Aliases: |
AER61, AOS4, C3orf64, EOGT1 |
Location: |
Chr.3:68975212-69013961 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EOGT | NM_001278689.1 | NP_001265618.1 | ||
| NM_173654.2 | NP_775925.1 | |||
| NR_103826.1 | ||||
| XM_005264743.2 | XP_005264800.1 | |||
| XM_011533599.1 | XP_011531901.1 | |||
| XM_011533600.1 | XP_011531902.1 | |||
| XM_011533602.1 | XP_011531904.1 | |||
| XM_011533603.1 | XP_011531905.1 | |||
| XM_017006204.1 | XP_016861693.1 | |||
| XM_017006205.1 | XP_016861694.1 | |||
| XM_017006206.1 | XP_016861695.1 | |||
| XM_017006207.1 | XP_016861696.1 | |||
| XM_017006208.1 | XP_016861697.1 | |||
| XM_017006209.1 | XP_016861698.1 | |||
| AJ868234.1 | CAI30569.1 | |||
| AK091089.1 | ||||
| AK126187.1 | BAC86479.1 | |||
| AK290356.1 | ||||
| AK294101.1 | ||||
| AK304102.1 | ||||
| BC060887.1 | AAH60887.1 | |||
| BX640821.1 | CAE45897.2 | |||
| BX648818.1 | ||||
| KC347596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3369576 | Chr.3:68996111 - 68997209 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map