Assay Details
Target Gene Details
Entrez Gene ID: | 222553 |
Gene Name: | solute carrier family 35 member F1 |
Gene Aliases: |
C6orf169, dJ230I3.1 |
Location: |
Chr.6:117907226-118317676 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC35F1 | NM_001029858.3 | NP_001025029.2 | ||
XM_005266865.4 | XP_005266922.1 | |||
AK295860.1 | ||||
BC028615.1 | AAH28615.1 | |||
BC036493.1 | AAH36493.1 | |||
BC114925.1 | AAI14926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3610502 | Chr.6:117920616 - 117923857 on Build GRCh38 | Loss |
![]() |
esv2673331 | Chr.6:117920543 - 117923961 on Build GRCh38 | Deletion |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
