Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFIX | NM_002501.3 | NP_002492.2 | ||
| XM_005259918.4 | XP_005259975.1 | |||
| XM_017026837.1 | XP_016882326.1 | |||
| BF115883.1 | ||||
| BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain |
|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss |
|
| nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map