Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIX | NM_002501.3 | NP_002492.2 | ||
XM_005259918.4 | XP_005259975.1 | |||
XM_017026837.1 | XP_016882326.1 | |||
BF115883.1 | ||||
BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain |
![]() |
nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion |
![]() |
nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
