Assay Details
Target Gene Details
Entrez Gene ID: | 66875 |
Gene Name: | SWT1 RNA endoribonuclease homolog (S. cerevisiae) |
Gene Aliases: |
1200016B10Rik, C1orf26 |
Location: |
Chr.1:151367698-151428450 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Swt1 | NM_025819.4 | NP_080095.3 | ||
AK004803.2 | BAB23575.2 | |||
AK013686.1 | BAB28953.1 | |||
BC028449.1 | ||||
BC055064.1 | AAH55064.1 | |||
BC060204.1 | AAH60204.1 | |||
BY062718.1 |
