Assay Details
Target Gene Details
Entrez Gene ID: | 66875 |
Gene Name: | SWT1 RNA endoribonuclease homolog (S. cerevisiae) |
Gene Aliases: |
1200016B10Rik, C1orf26 |
Location: |
Chr.1:151367698-151428450 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Swt1 | NM_025819.4 | 10 | 1552 | NP_080095.3 |
AK004803.2 | 10 | 1499 | BAB23575.2 | |
BC055064.1 | 10 | 1553 | AAH55064.1 | |
BC060204.1 | 10 | 1492 | AAH60204.1 |
