Assay Details
Target Gene Details
Entrez Gene ID: | 407243 |
Gene Name: | transmembrane protein 189 |
Gene Aliases: |
AI840826, Kua |
Location: |
Chr.2:167643225-167661544 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem189 | NM_145538.2 | NP_663513.1 | ||
AK088196.1 | BAC40204.1 | |||
AK151705.1 | ||||
AK213753.1 | ||||
BC002270.1 | AAH02270.1 |
