Assay Details
Target Gene Details
Entrez Gene ID: | 68259 |
Gene Name: | intraflagellar transport 80 |
Gene Aliases: |
4921524P20Rik, Wdr56, mKIAA1374 |
Location: |
Chr.3:68892499-69004653 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ift80 | NM_026641.2 | 15 | 1778 | NP_080917.1 |
AK019542.1 | 2 | 1695 | ||
AK042231.1 | 15 | 1703 | BAC31200.1 | |
AK129339.1 | 13 | 2026 | BAC98149.1 | |
AK147305.1 | 14 | 5305 | ||
BC013814.1 | 10 | 1067 | AAH13814.1 | |
BC034101.1 | 15 | 1673 | AAH34101.1 |
