Assay Details
Target Gene Details
Entrez Gene ID: | 13684 |
Gene Name: | eukaryotic translation initiation factor 4E |
Gene Aliases: |
EG668879, Eif4e-ps, If4e, eIF-4E |
Location: |
Chr.3:138526191-138557599 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4e | NM_007917.3 | NP_031943.3 | ||
| AK082867.1 | BAC38660.1 | |||
| AK146757.1 | ||||
| AK167097.1 | BAE39250.1 | |||
| BC010759.1 | AAH10759.1 | |||
| BC085087.1 | AAH85087.1 | |||
| M61731.1 | AAA37545.1 |
More Information
Set Membership: |
|

Genomic Map