Assay Details
Target Gene Details
Entrez Gene ID: | 12733 |
Gene Name: | chloride channel Ka |
Gene Aliases: |
C75963, CLC-K1, Clcnk1 |
Location: |
Chr.4:141384611-141398706 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clcnka | NM_001146307.1 | NP_001139779.1 | ||
| NM_024412.3 | NP_077723.3 | |||
| AF124848.1 | AAD21083.1 | |||
| AY373832.1 | AAQ81628.1 | |||
| BC037077.1 | AAH37077.1 |
More Information
Set Membership: |
|

Genomic Map