Assay Details
Target Gene Details
Entrez Gene ID: | 22044 |
Gene Name: | thyrotropin releasing hormone |
Gene Aliases: |
Pro-TRH, Trf |
Location: |
Chr.6:92242061-92244650 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trh | NM_009426.3 | NP_033452.2 | ||
| AK010666.1 | ||||
| BC053493.1 | AAH53493.1 | |||
| CD562178.1 | ||||
| CF951860.1 |
More Information
Set Membership: |
|

Genomic Map