Assay Details
Target Gene Details
Entrez Gene ID: | 320162 |
Gene Name: | centrosomal protein 95 |
Gene Aliases: |
4732496G21Rik, AI118346, AI448335, Ccdc45, D330027A14Rik, F630025I20Rik |
Location: |
Chr.11:106789252-106818861 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep95 | NM_001166685.1 | NP_001160157.1 | ||
| NM_177088.3 | NP_796062.2 | |||
| AK052705.1 | BAC35107.1 | |||
| AK076400.1 | BAC36324.1 | |||
| AK165351.1 | 2 | 234 | ||
| BC116308.1 | ||||
| BU695983.1 |

Genomic Map