Assay Details
Target Gene Details
Entrez Gene ID: | 387565 |
Gene Name: | CD300C antigen |
Gene Aliases: |
Clm6 |
Location: |
Chr.11:114956278-114960417 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd300c | NM_199225.1 | NP_954695.1 | ||
| AY457052.1 | AAR27943.1 |
More Information
Set Membership: |
|

Genomic Map