Assay Details
Target Gene Details
Entrez Gene ID: | 320162 |
Gene Name: | centrosomal protein 95 |
Gene Aliases: |
4732496G21Rik, AI118346, AI448335, Ccdc45, D330027A14Rik, F630025I20Rik |
Location: |
Chr.11:106789252-106818861 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cep95 | NM_001166685.1 | 7 | 743 | NP_001160157.1 |
NM_177088.3 | 8 | 872 | NP_796062.2 | |
AK052705.1 | 8 | 800 | BAC35107.1 | |
AK076400.1 | 8 | 850 | BAC36324.1 | |
BC116308.1 | 7 | 709 |
