Assay Details
Target Gene Details
Entrez Gene ID: | 108645 |
Gene Name: | methionine adenosyltransferase II, beta |
Gene Aliases: |
1110064C04Rik, 2410018D16Rik, AI182287, AU022853, MAT-II, MATIIbeta, TGR |
Location: |
Chr.11:40679314-40695203 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mat2b | NM_001199274.1 | NP_001186203.1 | ||
| NM_134017.2 | NP_598778.1 | |||
| AB070267.1 | BAD06938.1 | |||
| AK075600.1 | ||||
| AK075947.1 | BAC36076.1 | |||
| AK153126.1 | ||||
| AK162087.1 | BAE36716.1 | |||
| BC003457.1 | AAH03457.1 |
More Information
Set Membership: |
|

Genomic Map