Assay Details
Target Gene Details
Entrez Gene ID: | 16499 |
Gene Name: | potassium voltage-gated channel, shaker-related subfamily, beta member 3 |
Gene Aliases: |
C330022D06Rik, Kcnab4, mKv(beta)4 |
Location: |
Chr.11:69326258-69333041 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnab3 | NM_010599.4 | NP_034729.3 | ||
| AK029750.1 | ||||
| AK031806.1 | BAC27560.1 | |||
| AK044102.1 | BAC31778.1 | |||
| AK046194.1 | BAC32630.1 | |||
| AK049316.1 | BAC33678.1 | |||
| AK083134.1 | BAC38777.1 | |||
| BC017518.1 | AAH17518.1 | |||
| U65593.1 | AAB37264.1 |

Genomic Map