Assay Details
Target Gene Details
Entrez Gene ID: | 71770 |
Gene Name: | adaptor-related protein complex 2, beta 1 subunit |
Gene Aliases: |
1300012O03Rik, AI788979 |
Location: |
Chr.11:83298969-83405035 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap2b1 |
Target Gene Details
Entrez Gene ID: | 103737 |
Gene Name: | peroxisomal biogenesis factor 12 |
Gene Aliases: |
AI451906 |
Location: |
Chr.11:83294645-83298996 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pex12 | NM_134025.3 | NP_598786.1 | ||
| AK035552.1 | BAC29102.1 |
More Information
Set Membership: |
|

Genomic Map