Assay Details
Target Gene Details
Entrez Gene ID: | 19012 |
Gene Name: | phosphatidic acid phosphatase type 2A |
Gene Aliases: |
Hic53, Hpic53, LPP-1, LPP1, PAP-2a, PAP2-alpha, PAP2a, mPAP |
Location: |
Chr.13:112800727-112867894 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppap2a | NM_008247.3 | NP_032273.1 | ||
| NM_008903.2 | NP_032929.1 | |||
| AK077275.1 | BAC36724.1 | |||
| AK190923.1 | ||||
| AK195536.1 | ||||
| AK203135.1 | ||||
| CD742481.1 | ||||
| L43371.1 | AAA85353.1 |
Target Gene Details
Entrez Gene ID: | 72198 |
Gene Name: | superkiller viralicidic activity 2-like 2 (S. cerevisiae) |
Gene Aliases: |
2610528A15Rik, mKIAA0052 |
Location: |
Chr.13:112867780-112927380 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skiv2l2 | NM_028151.2 | 27 | 3206 | NP_082427.1 |
| AK012158.1 | 27 | 3206 | BAB28066.1 | |
| AK088426.1 | 26 | 3043 | BAC40348.1 | |
| AK129045.1 | 19 | 4329 | BAC97855.2 | |
| BC014810.1 | 19 | 2240 | AAH14810.1 | |
| BC029230.1 | 27 | 3196 | AAH29230.1 |
More Information
Set Membership: |
|

Genomic Map