Assay Details
Target Gene Details
Entrez Gene ID: | 223825 |
Gene Name: | maestro heat-like repeat family member 2B |
Gene Aliases: |
4930455B06Rik, Heatr7b2, SPIF |
Location: |
Chr.15:4898234-4962207 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mroh2b | NM_001166066.1 | NP_001159538.1 | ||
| KF778378.1 |
More Information
Set Membership: |
|

Genomic Map