Assay Details
Target Gene Details
Entrez Gene ID: | 17305 |
Gene Name: | MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase |
Gene Aliases: |
AW546563 |
Location: |
Chr.15:78755882-78773516 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfng | NM_008595.2 | NP_032621.1 | ||
| AF015769.1 | AAB71669.1 | |||
| AK143523.1 | BAE25415.1 | |||
| BC010983.1 | AAH10983.1 | |||
| U94349.1 | AAC53260.1 |
More Information
Set Membership: |
|

Genomic Map