Assay Details
Target Gene Details
Entrez Gene ID: | 12326 |
Gene Name: | calcium/calmodulin-dependent protein kinase IV |
Gene Aliases: |
A430110E23Rik, AI666733, CaMKIV, CaMKIV/Gr, D18Bwg0362e |
Location: |
Chr.18:32934957-33195767 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk4 | NM_009793.3 | NP_033923.2 | ||
| AK030219.1 | BAC26850.1 | |||
| AK043114.1 | BAC31462.1 | |||
| BC070420.1 | AAH70420.1 | |||
| X58995.1 | CAA41741.1 |
More Information
Set Membership: |
|

Genomic Map