Assay Details
Target Gene Details
Entrez Gene ID: | 68323 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 22 |
Gene Aliases: |
0610006K04Rik, AI851793, AW545547 |
Location: |
Chr.19:6993019-6996115 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt22 |
Target Gene Details
Entrez Gene ID: | 107328 |
Gene Name: | tRNA phosphotransferase 1 |
Gene Aliases: |
AU079016, AW045591, Tpt1, Tpt1h |
Location: |
Chr.19:6996131-6999048 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpt1 | NM_153597.2 | NP_705825.2 | ||
| NR_027351.1 | ||||
| AK038830.1 |
More Information
Set Membership: |
|

Genomic Map