Assay Details
Target Gene Details
Entrez Gene ID: | 23874 |
Gene Name: | phenylalanyl-tRNA synthetase, beta subunit |
Gene Aliases: |
C76708, Farsa, Farsl, Farslb, Frsb |
Location: |
Chr.1:78417958-78488897 on Build GRCm38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Farsb | NM_001278075.1 | NP_001265004.1 | ||
| AK012154.1 | BAB28064.1 | |||
| AK187456.1 |
Target Gene Details
Entrez Gene ID: | 433323 |
Gene Name: | sphingosine-1-phosphate phosphotase 2 |
Gene Aliases: |
SPPase2, Spp2 |
Location: |
Chr.1:78309501-78420289 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgpp2 | NM_001004173.2 | 5 | 2295 | NP_001004173.1 |
| AK081023.1 | 1 | 2151 | ||
| BC049995.1 | 5 | 2296 | AAH49995.1 |
More Information
Set Membership: |
|

Genomic Map