Assay Details
Target Gene Details
Entrez Gene ID: | 72699 |
Gene Name: | Lck interacting transmembrane adaptor 1 |
Gene Aliases: |
2810038K19Rik, LIME |
Location: |
Chr.2:181381235-181383628 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lime1 | NM_023684.2 | 5 | 1644 | NP_076173.1 |
| AF115339.1 | 5 | 1644 | AAG35210.1 | |
| AK012881.1 | 5 | 708 | ||
| AK172632.1 | 5 | 590 | BAE43105.1 | |
| BC023065.1 | 5 | 988 | AAH23065.1 |
Target Gene Details
Entrez Gene ID: | 72147 |
Gene Name: | zinc finger and BTB domain containing 46 |
Gene Aliases: |
2610019F01Rik, 4933406L05Rik, BZEL, Btbd4 |
Location: |
Chr.2:181371261-181492982 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb46 |
More Information
Set Membership: |
|

Genomic Map