Assay Details
Target Gene Details
Entrez Gene ID: | 69655 |
Gene Name: | CD164 sialomucin-like 2 |
Gene Aliases: |
2310047N01Rik |
Location: |
Chr.4:133220809-133224554 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd164l2 | NM_027152.1 | 6 | 830 | NP_081428.1 |
| AK009888.1 | 6 | 756 | BAB26564.1 | |
| AK139809.1 | 6 | 832 | ||
| AV057800.1 | 1 | 87 | ||
| BC111824.1 | AAI11825.1 |

Genomic Map