Assay Details
Target Gene Details
Entrez Gene ID: | 218194 |
Gene Name: | phosphatase and actin regulator 1 |
Gene Aliases: |
9630030F18Rik, Rpel1 |
Location: |
Chr.13:42680571-43138526 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phactr1 | NM_001005740.1 | 13 | 4747 | NP_001005740.1 |
| NM_001005748.2 | 5 | 3289 | NP_001005748.1 | |
| AK161124.1 | 1 | 2091 | ||
| BC061691.1 | 13 | 4747 | AAH61691.1 |
More Information
Set Membership: |
|

Genomic Map