Assay Details
Target Gene Details
Gene Symbol: | NeoR |
Gene Name: | Escherichia coli transposon Tn5 aminoglycoside-3'-O-phosphotransferase |
Gene Aliases: |
aph, Neo |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gene Symbol: | NeoR |
Gene Name: | Escherichia coli transposon Tn5 aminoglycoside-3'-O-phosphotransferase |
Gene Aliases: |
aph, Neo |